missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Rosiglitazone
Description
Rosiglitazone is a potent and selective PPARγ agonist (EC50 = 60 nM); exhibits no activity at PPARα and PPARβ. Promotes differentiation of pluripotent C3H10T1/2 stem cells into adipocytes. Also promotes differentiation of urothelial organoids in combination with Erlotinib (Cat. No. 7194). Exhibits antihyperglycemic activity in diabetic ob/ob mouse model. Antidiabetic agent.
Specifications
Specifications
| Chemical Name or Material | 5-[[4-[2-(Methyl-2-pyridinylamino)ethoxy]phenyl]methyl]-2,4-thiazolidinedione |
| CAS | 122320-73-4 |
| Synonym | BRL 49653 |
| Quantity | 10 mg |
| Target | PPAR gamma Receptor Agonists |
| Molecular Formula | C18H19N3O3S |
| Formula Weight | 357.43 |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?