missing translation for 'onlineSavingsMsg'
Learn More

Rose Bengal Sodium Salt, Technical Grade, TRC

Product Code. 30425115
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30425115 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30425115 Supplier Toronto Research Chemicals Supplier No. TRCR68490050G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acid Red 94
CAS 632-69-9
Molecular Formula C20 H2 Cl4 I4 O5 . 2 Na
Quantity 50 g
Synonym Fluorescein','4,5,6,7-tetrachloro-2',4',5',7'-tetraiodo-','disodium salt (8CI), Spiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one','4,5,6,7-tetrachloro-3',6'-dihydroxy-2',4',5',7'-tetraiodo-','disodium salt (9CI), 2',4',5',7'-Tetraiodo-3,4,5,6-tetrachlorofluorescein disodium salt, 3',4',5',6'-Tetrachloro-2,4,5,7-tetraiodofluorescein disodium salt, Acid red 94, Japan Red 105-1, Red No. 105-1, Rose Bengal Extra, Rose Bengal disodium, Rose Bengal disodium salt, Rose Bengal sodium, Rose Bengal sodium salt, Sodium tetraiodotetrachlorofluorescein
InChI Formula InChI=1S/C20H4Cl4I4O5.2Na/c21-9-7-8(10(22)12(24)11(9)23)20(33-19(7)31)3-1-5(25)15(29)13(27)17(3)32-18-4(20)2-6(26)16(30)14(18)28;;/h1-2,29-30H;;/q;2*+1/p-2
SMILES [Na+].[Na+].[O-]c1c(I)cc2c(Oc3c(I)c([O-])c(I)cc3C24OC(=O)c5c(Cl)c(Cl)c(Cl)c(Cl)c45)c1I
IUPAC Name disodium;4,5,6,7-tetrachloro-2',4',5',7'-tetraiodo-3-oxospiro[2-benzofuran-1,9'-xanthene]-3',6'-diolate
Molecular Weight (g/mol) 1017.64
Formula Weight 1015.463
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.