missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Rose Bengal, 85%, pure, high purity, Biological stain

Product Code. 13408438 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
missing translation for 'unitSize'
100g
1g
25g
500g
5g
This item is not returnable. View return policy

Product Code. 13408438

missing translation for 'mfr': Thermo Scientific Chemicals 189455000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 632-69-9 | C20H4Cl4I4Na2O5 | 1019.644 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 632-69-9
Molecular Formula C20H4Cl4I4Na2O5
Molecular Weight (g/mol) 1019.644
MDL Number MFCD00005043
InChI Key DPBQSRZQYAYDGY-UHFFFAOYSA-N
Synonym Acid Red 94, C.I. 45440
PubChem CID 87244310
IUPAC Name sodium;4,5,6,7-tetrachloro-3',6'-dihydroxy-2',4',5',7'-tetraiodospiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES C1=C2C(=C(C(=C1I)O)I)OC3=C(C(=C(C=C3C24C5=C(C(=C(C(=C5Cl)Cl)Cl)Cl)C(=O)O4)I)O)I.[Na].[Na]

Specifications

Absorption Ratio 1.21 to 1.95
Dye Content 80% min.
Assay Percent Range 85%
Solubility Information Solubility in water: soluble (36%).
Infrared Spectrum Authentic
Lambda Max 540 to 550nm (in water)
Formula Weight 1017.65
Loss on Drying 10% max. (105°C)
Percent Purity 85%
Grade Pure
Packaging Glass bottle
Color Bordeaux/Red to Brown/Red
Quantity 500 g
Flash Point >92°C
Physical Form Crystalline Powder
Chemical Name or Material Rose Bengal
Show More Show Less
compliance-icons
Product Identifier
  • Rose Bengal
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.