missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Rose Bengal

Product Code. 11448916
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448916 500 g 500g
11428916 25 g 25g
11438916 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11448916 Supplier Thermo Scientific Chemicals Supplier No. A17053.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 632-69-9 | C20H4Cl4I4Na2O5 | 1019.644 g/mol

Rose Bengal is a stain used in the eye drops to identify the eye damage. It finds application in the preparation of foraminifera for microscopic analysis, photosensitizing reactions and as a dye. In synthetic chemistry, it is utilized to convert triplet oxygen to singlet oxygen, which is used in [2+2] cycloadditions with alkenes. Its derivatives have important medical functions and used for the treatment of cancer. It acts as an alternative to phloxine B in Kreyberg's stain for keratin and mucus. As a fluorescent probe, it is used for the determination of NAD binding sites of alcohol dehydrogenase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 632-69-9
Molecular Formula C20H4Cl4I4Na2O5
Molecular Weight (g/mol) 1019.644
MDL Number MFCD00151169
InChI Key DPBQSRZQYAYDGY-UHFFFAOYSA-N
Synonym rose bengal disodium salt, r105 sodium, rose bengal sodium, rose-bengal 131 i natrium, food red no. 105, sodium salt, food red color no. 105, sodium salt, rosa bengala sodica 131 i inn-spanish, rose bengale 131 i sodique inn-french, sel disodique de rose bengale iodee 131 i, roseum bengalense 131 i natricum inn-latin
PubChem CID 87244310
IUPAC Name sodium;4,5,6,7-tetrachloro-3',6'-dihydroxy-2',4',5',7'-tetraiodospiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES C1=C2C(=C(C(=C1I)O)I)OC3=C(C(=C(C=C3C24C5=C(C(=C(C(=C5Cl)Cl)Cl)Cl)C(=O)O4)I)O)I.[Na].[Na]

Specifications

Melting Point >300°C
Quantity 500 g
Beilstein 3645857
Sensitivity Light sensitive
Merck Index 14,8262
Solubility Information Soluble in water.
Formula Weight 1017.65
Chemical Name or Material Rose Bengal
compliance-icons
Product Identifier
  • Rose Bengal
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.