missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Roscovitine

Click to view available options
Quantity:
10 mg
50mg
Unit Size:
10mg
50mg
Description
Cyclin-dependent kinase (cdk) inhibitor. Displays sub-micromolar inhibition of cdk1, cdk2, cdk5, cdk7 and cdk9 (reported IC50 values range from 0.16 - 0.28 μM for cdk5 to 0.8 μM for cdk7). Selective for cdks over a wide range of related kinases including ERK1 and ERK2. Arrests L1210 cells in G1 phase. Inhibits phosphorylation of vimentin in vivo. Antimitotic.
Specifications
Specifications
| CAS | 186692-46-6 |
| Synonym | roscovitine, seliciclib, r-roscovitine, rosco, 2-r-1-ethyl-2-hydroxyethylamino-6-benzylamino-9-isopropylpurine, unii-0es1c2kq94, insolution™ roscovitine, chembl14762, bmk1-e12 |
| Quantity | 10 mg |
| Molecular Formula | C19H26N6O |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction