missing translation for 'onlineSavingsMsg'
Learn More

Rosabulin, MedChemExpress

Product Code. 16416268
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16416268 10 mg 10mg
16498847 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16416268 Supplier Medchem Express Supplier No. HY14934

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Rosabulin (STA 5312) is a potent and orally active microtubule inhibitor that inhibits microtubule assembly. Rosabulin has broad-spectrum anti-tumor activity.

Rosabulin (STA 5312) is a potent and orally active microtubule inhibitor that inhibits microtubule assembly. Rosabulin has broad-spectrum anti-tumor activity.

Specifications

Chemical Name or Material Rosabulin
Physical Form Solid
CAS 501948-05-6
Color Green Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C22H16N4O2S
For Use With (Application) Cancer-programmed cell death
Synonym STA 5312
Solubility Information DMSO : 50 mg/mL (124.86 mM; Need ultrasonic)
SMILES O=C(NC1=CC(C)=NS1)C(C2=C3C=CC=CN3C(CC4=CC=C(C#N)C=C4)=C2)=O
Molecular Weight (g/mol) 400.45
Formula Weight 400.45
Percent Purity 98.72%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.