missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rosabulin, MedChemExpress
Description
Rosabulin (STA 5312) is a potent and orally active microtubule inhibitor that inhibits microtubule assembly. Rosabulin has broad-spectrum anti-tumor activity.
Specifications
Specifications
| Chemical Name or Material | Rosabulin |
| Physical Form | Solid |
| CAS | 501948-05-6 |
| Color | Green Yellow |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C22H16N4O2S |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | STA 5312 |
| Solubility Information | DMSO : 50 mg/mL (124.86 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?