missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RORB Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534162
This item is not returnable.
View return policy
Description
RORB Polyclonal Antibody for IHC (P)
Retinoid-related orphan receptor beta (ROR beta), a NR1 thyroid hormone receptor, affects the development and functioning of the central nervous system, including sensory input integration and circadian timing. Disruption of ROR beta in mice results in juvenile ataxia, retinal degeneration, circadian activity abnormalities, and delayed onset of male fertility. ROR beta binds to the monomeric response elements containing the sequence AnnTAGGTCA as a monomer. An alternatively spliced isoform, ROR beta2, has been isolated from rats, and the expression of ROR beta2 is confined to the pineal gland and retina. ROR beta2 shares common DNA- and putative ligand-binding domains with wild-type ROR beta but is characterized by a different amino-terminal domain. ROR beta expression has been documented in various regions of the mouse brain.
Specifications
| RORB | |
| Polyclonal | |
| Unconjugated | |
| RORB | |
| bA133M9.1; Nr1f2; Nuclear receptor ROR-beta; nuclear receptor RZR-beta; Nuclear receptor subfamily 1 group F member 2; RAR related orphan receptor B; RAR-related orphan receptor B; RAR-related orphan receptor beta; retinoic acid-binding receptor beta; retinoid-related orphan receptor b; retinoid-related orphan receptor beta; retinoid-related orphan receptor-beta; ror beta; Rorb; Rorbeta; ROR-BETA; rzrb; RZR-beta; sb:eu943; transcription factor; Unknown (protein for IMAGE:7173671) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100049831, 100169935, 100345295, 100771338, 225998, 309288, 395980, 484164, 528382, 6096 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P45446, Q8R1B8, Q92753 | |
| RORB | |
| Synthetic 18 amino acid peptide from hinge domain of human ROR Beta. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Sheep, Hamster, Horse, Chicken, Bat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction