missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RORA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10616-1-AP-150UL
This item is not returnable.
View return policy
Description
RORA is a receptor for retinoic acid belonging to the NR1 subfamily of nuclear hormone receptors with a nuclear receptor DNA binding domain. RORA binds either as a monomer or as a homodimer to the retinoic acid response element and thus regulates gene expression and also controls cell function. Though the exact function of RORA is yet unknown, it interacts with NM23-2, a nucleoside diphosphate kinase involved in organogenesis and differentiation, NM23-1, the product of a tumor metastasis suppressor candidate gene, CDK7, NCOA3 and NCOA6 coactivators. It is found to express in mesenchymal stem cells derived from bone marrow and are known to act by direct modulation of a bone matrix component. It is widely expressed in most of the tissues. Chromosomal aberrations involving RORA may be a cause of acute promyelocytic leukemia (APL).Specifications
| RORA | |
| Polyclonal | |
| Unconjugated | |
| RORA | |
| NR1F1, Nuclear receptor ROR alpha, Nuclear receptor RZR alpha, RAR related orphan receptor A, ROR1, ROR2, ROR3, RORA, RZR ALPHA, RZRA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19883, 300807, 6095 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P35398, P51448 | |
| RORA | |
| RORA Fusion Protein Ag0944 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction