missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROR2, Mouse anti-Human, Clone: 4B8, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00004920-M04.100ug
This item is not returnable.
View return policy
Description
Sequence: EVEVLDPNDPLGPLDGQDGPIPTLKGYFLNFLEPVNNITIVQGQTAILHCKVAGNPPPNVRWLKNDAPVVQEPRRIIIRKTEYGSRLRIQDLDTTDTGYYQCVATNGMKTSpecifications
| ROR2 | |
| Monoclonal | |
| Unconjugated | |
| receptor tyrosine kinase-like orphan receptor 2 | |
| BDB/BDB1/MGC163394/NTRKR2 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 4B8 | |
| In 1x PBS, pH 7.4 | |
| NM_004560 | |
| ROR2 | |
| ROR2 (NP_004551, 34 a.a. ∼ 143 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 4920 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction