missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ROR2 Monoclonal Antibody (CL5950)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531429
This item is not returnable.
View return policy
Description
Immunogen sequence: ADRAALLSEG ADDTQNAPE Highest antigen sequence indentity to the following orthologs: Rat - 79%, Mouse - 74%.
ROR2 (receptor tyrosine kinase-like orphan receptor 2) is a type I transmembrane protein that belongs to the ROR subfamily of cell surface receptors. It is thought to be involved in the early formation of the chondrocytes and cartilage and growth plate development. This Type I membrane protein is expressed at high levels during early embryonic development. The expression levels drop strongly around day 16 and there are only very low levels in adult tissues. Defects in ROR2 are a cause of brachydactyly type B1 (BDB1). BDB1 is an autosomal dominant skeletal disorder characterized by hypoplasia/aplasia of distal phalanges and nails. In BDB1 the middle phalanges are short but in addition the terminal phalanges are rudimentary or absent. Both fingers and toes are affected. The thumbs and big toes are usually deformed. Defects in ROR2 are a cause of recessive Robinow syndrome (RRS). RRS is an autosomal disorder characterized by skeletal dysplasia with generalized limb bone shortening, segmental defects of the spine, brachydactyly and a dysmorphic facial appearance. The protein contains 1 frizzled (FZ) domain, 1 immunoglobulin-like C2-type domain, and 1 kringle domain.
Specifications
| ROR2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q01974 | |
| ROR2 | |
| Synthetic peptide corresponding to Human ROR2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| CL5950 | |
| Unconjugated | |
| ROR2 | |
| BDB; BDB1; mRor2; neurotrophic tyrosine kinase receptor-related 2; neurotrophic tyrosine kinase, receptor related 2; neurotrophic tyrosine kinase, receptor-related 2; NTRKR2; receptor tyrosine kinase like orphan receptor 2; receptor tyrosine kinase-like orphan receptor 2; ROR2; Tyrosine-protein kinase transmembrane receptor ROR2 | |
| Mouse | |
| Protein A | |
| RUO | |
| 4920 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction