missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROR2 Antibody (CL5950), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61434
This item is not returnable.
View return policy
Description
ROR2 Monoclonal specifically detects ROR2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| ROR2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| ROR2 | |
| This antibody was developed against a synthetic peptide corresponding to amino acids: ADRAALLSEGADDTQNAPE | |
| 100 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| CL5950 | |
| Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 2-10 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| BDB, BDB1, EC 2.7.10.1, MGC163394, neurotrophic tyrosine kinase receptor-related 2, Neurotrophic tyrosine kinase, receptor-related 2, NTRKR2tyrosine-protein kinase transmembrane receptor ROR2, receptor tyrosine kinase-like orphan receptor 2 | |
| Mouse | |
| Protein A purified | |
| Protein Kinase, Wnt Signaling Pathway | |
| 4920 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction