Learn More
ROR2 Antibody (CL5950) [FITC], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61434F
Product Code. 30303487
  • £558.00 / 0.10mL
Description
ROR2 Monoclonal antibody specifically detects ROR2 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ROR2
Monoclonal
FITC
BDB, BDB1, EC 2.7.10.1, MGC163394, neurotrophic tyrosine kinase receptor-related 2, Neurotrophic tyrosine kinase, receptor-related 2, NTRKR2tyrosine-protein kinase transmembrane receptor ROR2, receptor tyrosine kinase-like orphan receptor 2
This antibody was developed using a synthetic peptide derived from Q01974, with the exact immunogen sequence remaining proprietary.
0.1 mL
Protein Kinase, Wnt Signaling Pathway
4920
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL5950
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified