missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROR1 Antibody (OTI3D11), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73931C
This item is not returnable.
View return policy
Description
ROR1 Monoclonal specifically detects ROR1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| ROR1 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ROR1 | |
| Full length recombinant protein of human ROR1 (NP_005003) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| OTI3D11 | |
| Western Blot | |
| EC 2.7.10.1, MGC99659, neurotrophic tyrosine kinase receptor-related 1, Neurotrophic tyrosine kinase, receptor-related 1, NTRKR1dJ537F10.1, receptor tyrosine kinase-like orphan receptor 1, tyrosine-protein kinase transmembrane receptor ROR1 | |
| Mouse | |
| Affinity Purified | |
| Protein Kinase | |
| 4919.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction