Learn More
ROR gamma/RORC/NR1F3 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Product Code. 18412648
Product Code. 18412648 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP224503MFV450
Rabbit Polyclonal Antibody
ROR gamma/RORC/NR1F3 Polyclonal antibody specifically detects ROR gamma/RORC/NR1F3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)