missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ropporin 1-like Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99172UV
This item is not returnable.
View return policy
Description
Ropporin 1-like Polyclonal specifically detects Ropporin 1-like in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Ropporin 1-like | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| AKAP-associated sperm protein, ASPFLJ23003, FLJ25776, radial spoke head 11 homolog, rhophilin associated tail protein 1-like, ROPN1-like protein, ropporin 1-like, ropporin-1-like protein, RSPH11 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human Ropporin 1-like. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 83853 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction