Learn More
Invitrogen™ ROPN1B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534792
Description
Recommended positive controls: A549, H1299, MCF-7. Predicted reactivity: Rhesus Monkey (97%). IHC notes, Requires antigen retrieval using heat mediated 10mM Citrate buffer (pH6.0) or Tris-EDTA buffer (pH8.0) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ROPN1B belongs to the ropporin family and contains 1 RIIa domain. ROPN1B interacts with RHPN1 and AKAP3. It may interact with SPA17.
Specifications
| ROPN1B | |
| Polyclonal | |
| Unconjugated | |
| ROPN1B | |
| AKAP-binding sperm protein ropporin; rhophilin associated tail protein 1B; rhophilin-associated protein 1B; ROPN1B; ropporin, rhophilin associated protein 1B; ropporin-1B; testis tissue sperm-binding protein Li 83P | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 152015 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9BZX4 | |
| ROPN1B | |
| Full length human ROPN1B Recombinant protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.