missing translation for 'onlineSavingsMsg'
Learn More

ROPN1 Antibody (4E11), Novus Biologicals™

Product Code. 18374349 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18374349 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18374349 Supplier Novus Biologicals Supplier No. H00054763M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ROPN1 Monoclonal antibody specifically detects ROPN1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ROPN1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4E11
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH15413
Gene Alias Cancer/testis antigen 91, CT91Rhophilin-associated protein 1A, ODF6RHPNAP1, outer dense fiber of sperm tails 6, rhophilin associated tail protein 1, ROPN1ADKFZp434B1222, ropporin, ropporin, rhophilin associated protein 1, ropporin-1A
Host Species Mouse
Immunogen ROPN1 (AAH15413, 1 a.a. ~ 120 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MWKVVNLPTDLFNSVMNVGRFTEEIEWLKFLALACSALGVTITKTLKIVCEVLSCDHNGGLPRIPFSTFQFLYTYIAEVDGEICASHVSRMLNYIEQEVIGPDGLITVNDFTQNPRVWLE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54763
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.