missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ropivacaine hydrochloride

Product Code. 17211302 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
17211302 250 mg 250mg
17201302 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 17211302

Brand: Thermo Scientific Chemicals 467222500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 98717-15-8 | C17H27ClN2O, C17H26N2O·HCl | 310.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98717-15-8
Molecular Formula C17H27ClN2O

Specifications

Quantity 250 mg
Melting Point 149.0°C to 151.0°C
Formula Weight 328.88
InChI Key NDNSIBYYUOEUSV-RSAXXLAASA-N
SMILES Cl.CCCN1CCCC[C@H]1C(=O)NC1=C(C)C=CC=C1C
IUPAC Name (2S)-N-(2,6-dimethylphenyl)-1-propylpiperidine-2-carboxamide hydrochloride
Molecular Weight (g/mol) 310.87
Chemical Name or Material Ropivacaine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.