missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ropinirole hydrochloride, MedChemExpress
Click to view available options
Quantity:
50 mg
100 mg
200 mg
Unit Size:
100mg
200mg
50mg
Description
Ropinirole (SKF 101468) hydrochloride is an orally active, potent D3/D2 receptor agonist with a Ki of 29 nM for D2 receptor. Ropinirole hydrochloride has pEC50s of 7.4, 8.4 and 6.8 for hD2, hD3 and hD4 receptors, respectively. Ropinirole hydrochloride has no affinity for the D1 receptors. Ropinirole hydrochloride has the potential for Parkinson's disease.
Specifications
Specifications
| Chemical Name or Material | Ropinirole hydrochloride |
| Physical Form | Solid |
| CAS | 91374-20-8 |
| Color | Yellow |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C16H25ClN2O |
| For Use With (Application) | Neuroscience-Neurodegeneration |
| Synonym | SKF 101468 hydrochloride |
| Solubility Information | H2O : 100 mg/mL (336.88 mM; Need ultrasonic) ∣DMSO : 16.67 mg/mL (56.16 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction