missing translation for 'onlineSavingsMsg'
Learn More

Ropinirole hydrochloride, MedChemExpress

Product Code. 16444427
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
200 mg
Unit Size:
100mg
200mg
50mg
This item is not returnable. View return policy

Product Code. 16444427

Brand: Medchem Express HYB0623A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ropinirole (SKF 101468) hydrochloride is an orally active, potent D3/D2 receptor agonist with a Ki of 29 nM for D2 receptor. Ropinirole hydrochloride has pEC50s of 7.4, 8.4 and 6.8 for hD2, hD3 and hD4 receptors, respectively. Ropinirole hydrochloride has no affinity for the D1 receptors. Ropinirole hydrochloride has the potential for Parkinson's disease.

Ropinirole (SKF 101468) hydrochloride is an orally active, potent D3/D2 receptor agonist with a Ki of 29 nM for D2 receptor. Ropinirole hydrochloride has pEC50s of 7.4, 8.4 and 6.8 for hD2, hD3 and hD4 receptors, respectively. Ropinirole hydrochloride has no affinity for the D1 receptors. Ropinirole hydrochloride has the potential for Parkinson's disease.

Specifications

Chemical Name or Material Ropinirole hydrochloride
Physical Form Solid
CAS 91374-20-8
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C16H25ClN2O
For Use With (Application) Neuroscience-Neurodegeneration
Synonym SKF 101468 hydrochloride
Solubility Information H2O : 100 mg/mL (336.88 mM; Need ultrasonic) ∣DMSO : 16.67 mg/mL (56.16 mM; Need ultrasonic)
SMILES O=C1NC2=C(C(CCN(CCC)CCC)=CC=C2)C1.Cl
Molecular Weight (g/mol) 296.84
Formula Weight 296.84
Percent Purity 99.25%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.