missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ropinirole hydrochloride

Product Code. 17211162 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 17211162

Brand: Thermo Scientific Chemicals 465002500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91374-20-8
Molecular Formula C16H25ClN2O

Specifications

Quantity 250 mg
Melting Point 241.0°C to 243.0°C
Formula Weight 296.84
InChI Key XDXHAEQXIBQUEZ-UHFFFAOYSA-N
SMILES [H+].[Cl-].CCCN(CCC)CCC1=CC=CC2=C1CC(=O)N2
IUPAC Name hydrogen 4-[2-(dipropylamino)ethyl]-2,3-dihydro-1H-indol-2-one chloride
Molecular Weight (g/mol) 296.84
Chemical Name or Material Ropinirole hydrochloride
compliance-icons compliance-icons
Product Identifier
  • Ropinirole hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P403-Store in a well-ventilated place.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Ropinirole hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.