missing translation for 'onlineSavingsMsg'
Learn More

Ronidazole-d3, TRC

Product Code. 30412802
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30412802 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30412802 Supplier Toronto Research Chemicals Supplier No. TRCR6402025MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ronidazole-d3
CAS 1015855-87-4
Purity Grade Notes HPLC
Molecular Formula C6 2H3 H5 N4 O4
Quantity 5 mg
Synonym Ronidazole D3, 1H-Imidazole-2-methanol','1-(methyl-d3)-5-nitro-','2-carbamate (ACI), 2-(Carbamoyloxymethyl)-1-methyl-d3-5-nitro-imidazole
InChI Formula InChI=1S/C6H8N4O4/c1-9-4(3-14-6(7)11)8-2-5(9)10(12)13/h2H,3H2,1H3,(H2,7,11)/i1D3
SMILES [2H]C([2H])([2H])n1c(COC(=O)N)ncc1[N+](=O)[O-]
IUPAC Name [5-nitro-1-(trideuteriomethyl)imidazol-2-yl]methyl carbamate
Molecular Weight (g/mol) 203.17
Formula Weight 203.0734
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.