missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RON Biosimilar Recombinant Monoclonal Antibody

Product Code. 30284316
Change view
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30284316 100 μg 100µg
30284300 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30284316 Supplier Invitrogen™ Supplier No. MA560083

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Monoclonal Antibody

For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term H5B14.

MST1R (RON) is a receptor tyrosine kinase involved in cell proliferation and motility. The protein is a membrane-spanning, disulfide-linked heterodimer, which results from cleavage of a glycosylated precursor into 35-kD (alpha) and 150-kD (beta) subunits. Ligand binding results in tyrosine phosphorylation of the beta chain. In knockout studies, MST1R/RON (-/-) mice failed to survive past the periimplantation period. The MST1R/RON gene has been mapped to 3p21, a region of frequent deletion or mutation in small cell lung and renal carcinoma, and has been implicated in the progression of several epithelial cancers. MST1R/Ron expression has been documented in many normal human tissues. ESTs have been isolated from several tissue libraries, including normal colon, mouth, prostate, and testis and cancerous colon, prostate, stomach, and uteru.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RON Biosimilar
Applications ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Classification Recombinant Monoclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 25 mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Host Species Human
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.