Learn More
Invitrogen™ Romosozumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA558772
 View more versions of this product
Product Code. 30283722
  • £1002.00 / 1mg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Romosozumab is a humanized monoclonal antibody indicated for the treatment of osteoporosis in postmenopausal women at high risk of fracture and patients who have failed in other treatments or are intolerant to other osteoporosis therapies9. Romosozumab prevents bone resorption and induces the formation of bone though it is associated with an increased risk of cardiac death, heart attack, and stroke in one study7, 8. In a comparison study of post menopausal women with osteoporosis and a past fracture, romosozumab for 12 months followed by alendronic acid for 12 months was superior to alendronic acid alone for 24 months3. Romosozumab also demonstrates a faster and larger increase in bone density than teriparatide4. Romosozumab is marketed in the United States by Amgen under the brand name Evinity7. Romosozumab was granted FDA approval on April 9, 2019.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Romosozumab Humanized
Recombinant Monoclonal
Unconjugated
785A070802; AMG 785; CDP-7851; romosozumab-aqqg; sclerostin Ab
Protein A
RUO
Human
Antibody
IgG2SA
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized