missing translation for 'onlineSavingsMsg'
Learn More

Romidepsin, MedChemExpress

Product Code. 16428917
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 16428917

Brand: Medchem Express HY15149

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Romidepsin (FK 228) is a Histone deacetylase (HDAC) inhibitor with anti-tumor activities. Romidepsin (FK 228) inhibits HDAC1, HDAC2, HDAC4, and HDAC6 with IC50s of 36 nM, 47 nM, 510 nM and 1.4 μM, respectively. Romidepsin (FK 228) is produced by Chromobacterium violaceum, induces cell G2/M phase arrest and apoptosis.

Romidepsin (FK 228) is a Histone deacetylase (HDAC) inhibitor with anti-tumor activities. Romidepsin (FK 228) inhibits HDAC1, HDAC2, HDAC4, and HDAC6 with IC50s of 36 nM, 47 nM, 510 nM and 1.4 μM, respectively. Romidepsin (FK 228) is produced by Chromobacterium violaceum, induces cell G2/M phase arrest and apoptosis.

Specifications

Chemical Name or Material Romidepsin
Physical Form Solid
CAS 128517-07-7
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C24H36N4O6S2
For Use With (Application) Cancer-programmed cell death
Synonym FK 228 FR 901228 NSC 630176
Solubility Information DMSO : ≥ 100 mg/mL (184.95 mM)
SMILES C/C=C(NC([C@@H](CSSCC/C=C/[C@@H](O1)CC2=O)NC([C@@H](C(C)C)N2)=O)=O)/C(N[C@@H](C(C)C)C1=O)=O
Molecular Weight (g/mol) 540.7
Formula Weight 540.7
Percent Purity 96.89%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.