missing translation for 'onlineSavingsMsg'
Learn More

Romidepsin, MedChemExpress

Product Code. 16451479
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16451479 1 mg 1mg
16428917 5 mg 5mg
16466328 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16451479 Supplier Medchem Express Supplier No. HY15149
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Romidepsin (FK 228) is a Histone deacetylase (HDAC) inhibitor with anti-tumor activities. Romidepsin (FK 228) inhibits HDAC1, HDAC2, HDAC4, and HDAC6 with IC50s of 36 nM, 47 nM, 510 nM and 1.4 μM, respectively. Romidepsin (FK 228) is produced by Chromobacterium violaceum, induces cell G2/M phase arrest and apoptosis.

Romidepsin (FK 228) is a Histone deacetylase (HDAC) inhibitor with anti-tumor activities. Romidepsin (FK 228) inhibits HDAC1, HDAC2, HDAC4, and HDAC6 with IC50s of 36 nM, 47 nM, 510 nM and 1.4 μM, respectively. Romidepsin (FK 228) is produced by Chromobacterium violaceum, induces cell G2/M phase arrest and apoptosis.

Specifications

Chemical Name or Material Romidepsin
Physical Form Solid
CAS 128517-07-7
Color White
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C24H36N4O6S2
For Use With (Application) Cancer-programmed cell death
Synonym FK 228 FR 901228 NSC 630176
Solubility Information DMSO : ≥ 100 mg/mL (184.95 mM)
SMILES C/C=C(NC([C@@H](CSSCC/C=C/[C@@H](O1)CC2=O)NC([C@@H](C(C)C)N2)=O)=O)/C(N[C@@H](C(C)C)C1=O)=O
Molecular Weight (g/mol) 540.7
Formula Weight 540.7
Percent Purity 96.89%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.