missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Roluperidone, MedChemExpress
Description
Roluperidone (CYR-101) is a novel cyclic amide derivative that has high equipotent affinities for 5-HT2A and sigma-2 receptors (Ki of 7.53 nM and 8.19 nM for 5-HT2A and sigma-2, respectively).
Specifications
Specifications
| Chemical Name or Material | Roluperidone |
| CAS | 359625-79-9 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C22H23FN2O2 |
| Quantity | 100 mg |
| Synonym | CYR-101 MIN-101 MT-210 |
| Solubility Information | DMSO : 33.33 mg/mL (90.96 mM; Need ultrasonic) |
| SMILES | O=C1N(CC2CCN(CC(C3=CC=C(F)C=C3)=O)CC2)CC4=C1C=CC=C4 |
| Molecular Weight (g/mol) | 366.43 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?