missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rofecoxib, MedChemExpress
Description
Rofecoxib is a potent, specific and orally active COX-2 inhibitor, with IC50s of 26 and 18 nM for human COX-2 in human osteosarcoma cells and Chinese hamster ovary cells, with a 1000-fold selectivity for COX-2 over human COX-1 (IC50 > 50 μM in U937 cells and > 15 μM in Chinese hamster ovary cells).
Specifications
Specifications
| Chemical Name or Material | Rofecoxib |
| CAS | 162011-90-7 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C17H14O4S |
| Quantity | 100 mg |
| Synonym | MK 966 |
| Solubility Information | DMSO : 33.33 mg/mL (106.02 mM; Need ultrasonic) |
| SMILES | O=C1OCC(C2=CC=C(S(=O)(C)=O)C=C2)=C1C3=CC=CC=C3 |
| Molecular Weight (g/mol) | 314.36 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?