missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rocuronium Bromide, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Description
Rocuronium Bromide (ORG 9426 Bromide) is an aminosteroid non-depolarizing neuromuscular blocker or muscle relaxant used in modern anaesthesia, to facilitate endotracheal intubation and to provide skeletal musclerelaxation during surgery or mechanical ventilation.
Specifications
Specifications
| Chemical Name or Material | Rocuronium Bromide |
| CAS | 119302-91-9 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C32H53BrN2O4 |
| Quantity | 100 mg |
| Synonym | ORG 9426 Bromide |
| Solubility Information | DMSO : 100 mg/mL (164.02 mM; Need ultrasonic) ∣H2O : 100 mg/mL (164.02 mM; Need ultrasonic) |
| SMILES | C[C@@]12[C@](C[C@H]([N+]3(CC=C)CCCC3)[C@@H]2OC(C)=O)([H])[C@]([C@]4([H])CC1)([H])CC[C@@]([C@]4(C)C5)([H])C[C@H](O)[C@H]5N6CCOCC6.[Br-] |
| Molecular Weight (g/mol) | 609.68 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction