missing translation for 'onlineSavingsMsg'
Learn More

Rocuronium Bromide, MedChemExpress

Product Code. 16423367
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16423367 50 mg 50mg
16417316 100 mg 100mg
16407348 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16423367 Supplier Medchem Express Supplier No. HY17440
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Rocuronium Bromide (ORG 9426 Bromide) is an aminosteroid non-depolarizing neuromuscular blocker or muscle relaxant used in modern anaesthesia, to facilitate endotracheal intubation and to provide skeletal musclerelaxation during surgery or mechanical ventilation.

Rocuronium Bromide (ORG 9426 Bromide) is an aminosteroid non-depolarizing neuromuscular blocker or muscle relaxant used in modern anaesthesia, to facilitate endotracheal intubation and to provide skeletal musclerelaxation during surgery or mechanical ventilation.

Specifications

Chemical Name or Material Rocuronium Bromide
CAS 119302-91-9
Color White
Purity Grade Notes Research
Molecular Formula C32H53BrN2O4
Quantity 50 mg
Synonym ORG 9426 Bromide
Solubility Information DMSO : 100 mg/mL (164.02 mM; Need ultrasonic) ∣H2O : 100 mg/mL (164.02 mM; Need ultrasonic)
SMILES C[C@@]12[C@](C[C@H]([N+]3(CC=C)CCCC3)[C@@H]2OC(C)=O)([H])[C@]([C@]4([H])CC1)([H])CC[C@@]([C@]4(C)C5)([H])C[C@H](O)[C@H]5N6CCOCC6.[Br-]
Molecular Weight (g/mol) 609.68
Formula Weight 609.68
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.