Learn More
Cole-Parmer™ Rocking Platform
Complement your hybridization incubation systems with this Rocking Platform.
Brand: Cole-Parmer™ 39455-16
Code : 04
Additional Details : Weight : 0.80000kg
Description
- Allows a number of hybridization tubes to be placed above.
- Manufactured from stainless steel.
- Enables membranes to be processed using a wave motion.
- Speed range of 0 to 60 oscillations per minute.
Specifications
HI-200 Series Hybridization Incubators | |
Stainless Steel | |
15 to 60 rpm |
Hybridization Tubes | |
Rocking Platform |