Learn More
Cole-Parmer™ Rocking Platform
Complement your hybridization incubation systems with this Rocking Platform.
Brand:  Cole-Parmer™ 39455-09
Code : A0
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11563839
  • £192.14 / Each
Estimated Shipment: 06-06-2024
to see stock.
Add to basket
Description
Description
  • Allows a number of hybridization tubes to be placed above.
  • Manufactured from stainless steel.
  • Enables membranes to be processed using a wave motion.
  • Speed range of 0 to 60 oscillations per minute.
Specifications
Specifications
HI-200 Series Hybridization Incubators
Stainless Steel
20 to 60 rpm
Hybridization Tubes
Rocking Platform