Learn More
SP Genevac™ Rocket Synergy 2 Dionex 250 mL ASE Bottle Adaptor
Product Code. 31723002
Change view
Click to view available options
Material:
Plastic
Unit Size:
Pack of 6
Product Code. Material Unit Size
31723002 Plastic Pack of 6
1 options
Product Code. 31723002 Supplier SP Genevac™ Supplier No. 702010
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Set of 6 adaptors for 250 mL Dionex™ bottles
  • Allows up to 6 bottles to be processed simultaneously
  • Set of 6
  • Adapter that allows the use of Dionex™ 250 mL ASE Collection Bottles
  • Compatible with Rocket Synergy 2 Rocket Synergy
  • Only compatible with Flask Rotors
Specifications
Material Plastic
Type Adapter Set