missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROCK2(middle) Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21645-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is specific to ROCK2.
The function of this protein remains unknown.Specifications
| ROCK2(middle) | |
| Polyclonal | |
| Unconjugated | |
| ROCK2 | |
| KIAA0619, p164 ROCK 2, Rho kinase 2, ROCK2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19878, 25537, 9475 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.52 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75116, P70336, Q62868 | |
| Rock2 | |
| ROCK2(middle) Fusion Protein Ag16330 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction