missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROCK1 Rabbit anti-Human, Mouse, Rat, Clone: H5E4, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £367.00
Specifications
| Antigen | ROCK1 |
|---|---|
| Clone | H5E4 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18319382
|
Bio-Techne
NBP3-15350-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18355026
|
Novus Biologicals
NBP3-15350-100UL |
100 μg |
£367.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ROCK1 Monoclonal antibody specifically detects ROCK1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| ROCK1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Breast Cancer, Cancer, Signal Transduction, Wnt Signaling Pathway | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 6093 | |
| IgG | |
| Affinity purified |
| H5E4 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 2.7.11, EC 2.7.11.1, MGC131603, MGC43611, p160 ROCK-1, p160ROCK, p160-ROCK, PRO0435, Renal carcinoma antigen NY-REN-35, Rho kinase, rho-associated protein kinase 1, Rho-associated, coiled-coil containing protein kinase 1, Rho-associated, coiled-coil-containing protein kinase 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1100-1200 of human ROCK1 (Q13464). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title