missing translation for 'onlineSavingsMsg'
Learn More

ROCK1 Mouse anti-Human, Mouse, Rat, Clone: 3G5E12, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66782-1-IG-150UL

 View more versions of this product

Product Code. 16838975

  • £349.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The Rho target protein, Rho-associated coiled coil-containing protein kinase (ROCK)-1 has been found to be a new Caspase-3 substrate. ROCK, one of the effectors of the small GTPase Rho, has recently been shown to contribute significantly to myosin light chain (MLC) activation through two pathways: direct phosphorylation of MLC and phosphorylation of MLC phosphatase, leading to its inhibition. The increase in cellular contractility that is necessary for apoptotic membrane blebbing implies sustained augmentation of MLC phosphorylation. ROCK-1 consists of an amino-terminal kinase domain and an inhibitory cysteine/histidine-rich C-terminal domain that is located within a pleckstrin-homology region. They are joined by a variable region that contains the Rho-binding domain. During apoptosis, ROCK-I is cleaved by caspase-3 at a conserved DETD1113/G sequence and its carboxy-terminal inhibitory domain is removed, resulting in deregulated and constitutive kinase activity. The caspase-3-mediated cleavage and activation of ROCK I induces phosphorylation of MLC and membrane blebbing, one of the first events in the execution phase of apoptosis.
Specifications

Specifications

ROCK1
Monoclonal
0.8 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P70335, Q13464, Q63644
ROCK1
ROCK1 Fusion Protein Ag16317
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG1
Western Blot
3G5E12
Unconjugated
ROCK1
p160 ROCK 1, P160ROCK, PRO0435, ROCK1
Mouse
Protein G
RUO
19877, 6093, 81762
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.