missing translation for 'onlineSavingsMsg'
Learn More

ROC-325, MedChemExpress

Product Code. 16406627
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16406627 5 mg 5mg
16473898 10 mg 10mg
16454936 100 mg 100mg
16410267 50 mg 50mg
16402906 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16406627 Supplier Medchem Express Supplier No. HY103706

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ROC-325 is a potent and orally active autophagy inhibitor with a strong anticancer activity. ROC-325 induces the deacidification of lysosomes, accumulation of autophagosomes, and disrupted autophagic flux. ROC-325 also induces renal cell carcinoma apoptosis.

ROC-325 is a potent and orally active autophagy inhibitor with a strong anticancer activity. ROC-325 induces the deacidification of lysosomes, accumulation of autophagosomes, and disrupted autophagic flux. ROC-325 also induces renal cell carcinoma apoptosis.

Specifications

Chemical Name or Material ROC-325
CAS 1859141-26-6
Color Orange
Purity Grade Notes Research
Molecular Formula C28H27ClN4OS
Quantity 5 mg
Solubility Information DMSO : 32 mg/mL (63.61 mM; Need ultrasonic) ∣H2O : 1 mg/mL (1.99 mM; Need ultrasonic)
SMILES O=C1C2=C(SC3=C1C=CC=C3)C(C)=CC=C2NCCN(CCNC4=CC=NC5=CC(Cl)=CC=C45)C
Molecular Weight (g/mol) 503.06
Formula Weight 503.06
Percent Purity 99.91%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.