missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ 18 Gauge Fill Tube

Product Code. 15747659
Click to view available options
Overall Length (Metric):
650 mm
This item is not returnable. View return policy

Product Code. 15747659

Brand: Hamilton™ 240010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Provides the perfect seal including minimized dead volume for 1/4 in.-28 UNF or M6 flat bottom valve ports.

  • Tubing assembly
  • For use with all Microlab™ 500 instruments
  • Tubing manufactured from FEP

Specifications

Description 18 gauge
Material FEP
Overall Length (Metric) 650 mm
For Use With Microlab 500 A, B, & C series
Packaging Quantity 1 Pack
Thread Size M6
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.