missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ROBO2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA582481

Product Code. 16339444

  • £480.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: PVNNSNSGPN EIGNFGRGDV LPPVPGQGDK TATMLSDGAI YSSIDFTTKT SYNSSSQITQ ATPYATTQIL HSNSIHELAV DLPDPQWKSS IQQKTDLMGF GYSLPDQNKG NNGGKGGKKK KNKNSSKPQK NNGSTWA Highest antigen sequence indentity to the following orthologs: Rat - 96%, Mouse - 97%.

ROBO2 belongs to the ROBO family, part of the immunoglobulin superfamily proteins that are highly conserved from fly to human. ROBO2 is a receptor for SLIT2, molecules known to function in axon guidance and cell migration. Defects in this gene are the cause of vesicoureteral reflux type 2. Alternatively spliced transcript variants encoding different isoforms have been described for ROBO2.This gene belongs to the ROBO family, part of the immunoglobulin superfamily proteins that are highly conserved from fly to human. The encoded protein is a receptor for SLIT2, molecules known to function in axon guidance and cell migration. Defects in this gene are the cause of vesicoureteral reflux type 2. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ROBO2
Polyclonal
Unconjugated
ROBO2
2600013A04Rik; 9430089E08Rik; ast; astray; BB097918; D230004I22Rik; KIAA1568; mKIAA1568; ROBO2; roundabout 2; roundabout guidance receptor 2; roundabout homolog 2; roundabout homolog 2 (Drosophila); roundabout homolog 2-like; roundabout, axon guidance receptor, homolog 2; roundabout, axon guidance receptor, homolog 2 (Drosophila); roundabout2; SAX3; sb:eu937; Unknown (protein for IMAGE:8049638); unp1881; zmp:0000000549
Rabbit
Antigen affinity chromatography
RUO
268902, 6092
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.10 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q7TPD3, Q9HCK4
ROBO2
Recombinant protein corresponding to Human ROBO2. Recombinant protein control fragment (Product #RP-90546).
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.