missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ROBO2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547461
This item is not returnable.
View return policy
Description
In direct ELISAs and Western blots, less than 2% cross-reactivity with recombinant rat ROBO1, recombinant human (rh) ROBO3 and rhROBO4 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
ROBO2 belongs to the ROBO family, part of the immunoglobulin superfamily proteins that are highly conserved from fly to human. ROBO2 is a receptor for SLIT2, molecules known to function in axon guidance and cell migration. Defects in this gene are the cause of vesicoureteral reflux type 2. Alternatively spliced transcript variants encoding different isoforms have been described for ROBO2.This gene belongs to the ROBO family, part of the immunoglobulin superfamily proteins that are highly conserved from fly to human. The encoded protein is a receptor for SLIT2, molecules known to function in axon guidance and cell migration. Defects in this gene are the cause of vesicoureteral reflux type 2. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Specifications
| ROBO2 | |
| Polyclonal | |
| Unconjugated | |
| ROBO2 | |
| 2600013A04Rik; 9430089E08Rik; ast; astray; BB097918; D230004I22Rik; KIAA1568; mKIAA1568; ROBO2; roundabout 2; roundabout guidance receptor 2; roundabout homolog 2; roundabout homolog 2 (Drosophila); roundabout homolog 2-like; roundabout, axon guidance receptor, homolog 2; roundabout, axon guidance receptor, homolog 2 (Drosophila); roundabout2; SAX3; sb:eu937; Unknown (protein for IMAGE:8049638); unp1881; zmp:0000000549 | |
| Goat | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6092 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q9HCK4 | |
| ROBO2 | |
| Mouse myeloma cell line NS0-derived recombinant human ROBO2 Ser22-Ala312, RPVA, Pro313-Pro859. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction