missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROBO1 Antibody (Asclepius Technology patent anti-Robo1 CAR) [Janelia Fluor™ 635], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28772JF635
This item is not returnable.
View return policy
Description
ROBO1 Monoclonal antibody specifically detects ROBO1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| ROBO1 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| DUTT1Drosophila) homolog 1, H-Robo-1, MGC131599, MGC133277, roundabout, axon guidance receptor, homolog 1 (Drosophila) | |
| ROBO1 | |
| 0.1 mL | |
| Angiogenesis, Cancer, Extracellular Matrix | |
| 6091 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Asclepius Technology patent anti-Robo1 CAR | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction