missing translation for 'onlineSavingsMsg'
Learn More

RO1138452, MedChemExpress

Product Code. 16495176
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16495176 100 mg 100mg
16470577 50 mg 50mg
16443006 25 mg 25mg
16436967 5 mg 5mg
16414248 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16495176 Supplier Medchem Express Supplier No. HY108912
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress RO1138452 is a potent and selective IP (prostacyclin) receptor antagonist. RO1138452 displays high affinity for IP receptors. In human platelets, pKi is 9.3±0.1; in a recombinant IP receptor system, pKi is 8.7±0.06.

RO1138452 is a potent and selective IP (prostacyclin) receptor antagonist. RO1138452 displays high affinity for IP receptors. In human platelets, pKi is 9.3±0.1

Specifications

Chemical Name or Material RO1138452
CAS 221529-58-4
Color Light Nude
Purity Grade Notes Research
Molecular Formula C19H23N3O
Quantity 100 mg
Synonym CAY10441
Solubility Information DMSO : 100 mg/mL (323.20 mM; Need ultrasonic) ∣Ethanol : 25 mg/mL (80.80 mM; Need ultrasonic)
SMILES CC(C)OC1=CC=C(CC2=CC=C(NC3=NCCN3)C=C2)C=C1
Molecular Weight (g/mol) 309.41
Formula Weight 309.41
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.