missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ro 67-7476, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
Ro 67-7476 is a potent positive allosteric modulator of mGluR1 and potentiates glutamate-induced calcium release in HEK293 cells expressing rat mGluR1a with an EC50 of 60.1 nM. Ro 67-7476 is a potent P-ERK1/2 agonist and activates ERK1/2 phosphorylation in the absence of exogenously added glutamate (EC50=163.3 nM).
Specifications
Specifications
| Chemical Name or Material | Ro 67-7476 |
| CAS | 298690-60-5 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C17H18FNO2S |
| Quantity | 100 mg |
| Solubility Information | DMSO : ≥ 40 mg/mL (125.24 mM) |
| SMILES | O=S(N1[C@H](C2=CC=C(F)C=C2)CCC1)(C3=CC=C(C)C=C3)=O |
| Molecular Weight (g/mol) | 319.39 |
| Formula Weight | 319.39 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction