missing translation for 'onlineSavingsMsg'
Learn More

Ro 64-5229, Tocris Bioscience™

Catalog Number p-4784436
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
246852-46-0
C17H19Cl2N3O
352.259
STCVFKBRXOEQRF-YBEGLDIGSA-N
d09ubs, 1-z-2-cycloheptyloxy-2-2,6-dichlorophenyl ethenyl-1,2,4-triazole, 1-z-beta-cycloheptyloxy-2,6-dichlorostyryl-1h-1,2,4-triazole, z-1-2-cycloheptyloxy-2-2,6-dichlorophenyl ethenyl-1h-1,2,4-triazole, 1-z-2-cycloheptyloxy-2-2,6-dichloro-phenyl-vinyl-1h-1,2,4 triazole, 1-2-cycloheptyloxy-2-2,6-dichloro-phenyl-vinyl-1h-1,2,4 triazole ro 64-5229
10428048
1-[(Z)-2-cycloheptyloxy-2-(2,6-dichlorophenyl)ethenyl]-1,2,4-triazole
C1CCCC(CC1)OC(=CN2C=NC=N2)C3=C(C=CC=C3Cl)Cl
This item is not returnable. View return policy
246852-46-0
C17H19Cl2N3O
352.259
STCVFKBRXOEQRF-YBEGLDIGSA-N
d09ubs, 1-z-2-cycloheptyloxy-2-2,6-dichlorophenyl ethenyl-1,2,4-triazole, 1-z-beta-cycloheptyloxy-2,6-dichlorostyryl-1h-1,2,4-triazole, z-1-2-cycloheptyloxy-2-2,6-dichlorophenyl ethenyl-1h-1,2,4-triazole, 1-z-2-cycloheptyloxy-2-2,6-dichloro-phenyl-vinyl-1h-1,2,4 triazole, 1-2-cycloheptyloxy-2-2,6-dichloro-phenyl-vinyl-1h-1,2,4 triazole ro 64-5229
10428048
1-[(Z)-2-cycloheptyloxy-2-(2,6-dichlorophenyl)ethenyl]-1,2,4-triazole
C1CCCC(CC1)OC(=CN2C=NC=N2)C3=C(C=CC=C3Cl)Cl

Selective, non-competitive mGlu2 antagonist

Selective, non-competitive mGlu2 antagonist. Inhibits GTPγ35S binding to mGlu2-containing membranes (IC50 = 0.11 μM).
Quantity 10 mg
Formula Weight 352.26
Percent Purity >99%
Chemical Name or Material Ro 64-5229
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'