missing translation for 'onlineSavingsMsg'
Learn More

Ro 5126766, MedChemExpress

Product Code. 16473577
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16473577 50 mg 50mg
16467558 10 mg 10mg
16467496 100 mg 100mg
16430038 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16473577 Supplier Medchem Express Supplier No. HY18652

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ro 5126766 (CH5126766) is a first-in-class dual MEK/RAF inhibitor that allosterically inhibits BRAFV600E, CRAF, MEK, and BRAF (IC50: 8.2, 56, 160 nM, and 190 nM, respectively).

Ro 5126766 (CH5126766) is a first-in-class dual MEK/RAF inhibitor that allosterically inhibits BRAFV600E, CRAF, MEK, and BRAF (IC50: 8.2, 56, 160 nM, and 190 nM, respectively).

Specifications

Chemical Name or Material Ro 5126766
CAS 946128-88-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H18FN5O5S
Quantity 50 mg
Synonym CH5126766
Solubility Information DMSO : 125 mg/mL (265.13 mM; Need ultrasonic)
SMILES O=S(NC)(NC1=NC=CC(CC2=C(C)C3=CC=C(OC4=NC=CC=N4)C=C3OC2=O)=C1F)=O
Molecular Weight (g/mol) 471.46
Formula Weight 471.46
Percent Purity 98.08%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.