missing translation for 'onlineSavingsMsg'
Learn More

Ro 31-8220 mesylate, MedChemExpress

Product Code. p-200099180
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16415898

Brand: Medchem Express HY13866

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ro 31-8220 mesylate is a potent PKC inhibitor, with IC50s of 5, 24, 14, 27, 24 and 23 nM for PKCα, PKCβI, PKCβII, PKCγ, PKCε and rat brain PKC, respectively. Ro 31-8220 also significantly inhibits MAPKAP-K1b, MSK1, S6K1 and GSK3β (IC50s, 3, 8, 15, and 38 nM, respectively), with no effect on MKK3, MKK4, MKK6 and MKK7.

Ro 31-8220 mesylate is a potent PKC inhibitor, with IC50s of 5, 24, 14, 27, 24 and 23 nM for PKCα, PKCβI, PKCβII, PKCγ, PKCε and rat brain PKC, respectively. Ro 31-8220 also significantly inhibits MAPKAP-K1b, MSK1, S6K1 and GSK3β (IC50s, 3, 8, 15, and 38 nM, respectively), with no effect on MKK3, MKK4, MKK6 and MKK7.

Specifications

Chemical Name or Material Ro 31-8220 mesylate
CAS 138489-18-6
Color Orange
Purity Grade Notes Research
Molecular Formula C26H27N5O5S2
Quantity 10 mg
Synonym Ro 31-8220 methanesulfonate Bisindolylmaleimide IX mesylate
Solubility Information DMSO : 35.71 mg/mL (64.50 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES NC(SCCCN1C=C(C2=C(C3=CN(C)C4=C3C=CC=C4)C(NC2=O)=O)C5=C1C=CC=C5)=N.CS(=O)(O)=O
Molecular Weight (g/mol) 553.65
Formula Weight 553.65
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.