missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ro 31-8220 mesylate, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Ro 31-8220 mesylate is a potent PKC inhibitor, with IC50s of 5, 24, 14, 27, 24 and 23 nM for PKCα, PKCβI, PKCβII, PKCγ, PKCε and rat brain PKC, respectively. Ro 31-8220 also significantly inhibits MAPKAP-K1b, MSK1, S6K1 and GSK3β (IC50s, 3, 8, 15, and 38 nM, respectively), with no effect on MKK3, MKK4, MKK6 and MKK7.
Specifications
Specifications
| Chemical Name or Material | Ro 31-8220 mesylate |
| CAS | 138489-18-6 |
| Color | Orange |
| Purity Grade Notes | Research |
| Molecular Formula | C26H27N5O5S2 |
| Quantity | 10 mg |
| Synonym | Ro 31-8220 methanesulfonate Bisindolylmaleimide IX mesylate |
| Solubility Information | DMSO : 35.71 mg/mL (64.50 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble) |
| SMILES | NC(SCCCN1C=C(C2=C(C3=CN(C)C4=C3C=CC=C4)C(NC2=O)=O)C5=C1C=CC=C5)=N.CS(=O)(O)=O |
| Molecular Weight (g/mol) | 553.65 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction