missing translation for 'onlineSavingsMsg'
Learn More

RO-28-1675, MedChemExpress

Product Code. 16456767
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16456767 5 mg 5mg
16434068 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16456767 Supplier Medchem Express Supplier No. HY10595
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress RO-28-1675 is a potent allosteric glucokinase (GK) activator with an EC50 of 54 nM. RO-28-1675 can be used for the research of type 2 diabetes.

RO-28-1675 is a potent allosteric glucokinase (GK) activator with an EC50 of 54 nM. RO-28-1675 can be used for the research of type 2 diabetes.

Specifications

Chemical Name or Material RO-28-1675
CAS 300353-13-3
Color White
Purity Grade Notes Research
Molecular Formula C18H22N2O3S2
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (132.10 mM; Need ultrasonic)
SMILES O=C([C@@H](C1=CC=C(C=C1)S(=O)(C)=O)CC2CCCC2)NC3=NC=CS3
Molecular Weight (g/mol) 378.51
Formula Weight 378.51
Percent Purity 99.62%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.