missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ro 25-6981 Maleate, MedChemExpress
Description
Ro 25-6981 Maleate is a potent, selective and activity-dependent NR2B subunit specific NMDA receptor antagonist. Ro 25-6981 Maleat shows anticonvulsant and anti-parkinsonian activity. Ro 25-6981 Maleate has the potential for the research of parkinson's disease (PD).
Specifications
Specifications
| Chemical Name or Material | Ro 25-6981 Maleate |
| CAS | 1312991-76-6 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C26H33NO6 |
| Quantity | 100 mg |
| Solubility Information | DMSO : 100 mg/mL (219.52 mM; Need ultrasonic) ∣H2O : 8.33 mg/mL (18.29 mM; ultrasonic and warming and heat to 45°C) |
| SMILES | OC1=CC=C([C@H](O)[C@@H](C)CN2CCC(CC3=CC=CC=C3)CC2)C=C1.O=C(O)/C=C\C(O)=O |
| Molecular Weight (g/mol) | 455.54 |
| Formula Weight | 455.54 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?